Anti-LYST Mouse Monoclonal Antibody [clone: 3H1]
Catalog # ABNOH00001130-M07
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:lysosomal trafficking regulator
- Antigen symbol:LYST
- Clonality:Monoclonal
- Clone:3H1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:1130
- Antigen synonyms:CHS1|CHS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LYST
Clonality: Monoclonal
Clone: 3H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human