Order Entry
Denmark
ContactUsLinkComponent
 
Anti-CDX2 Mouse Monoclonal Antibody [clone: 1C7]
Catalog # ABNOH00001045-M01
Supplier:  Abnova
undefined
Anti-CDX2 Mouse Monoclonal Antibody [clone: 1C7]
Catalog # ABNOH00001045-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Caudal type homeobox 2
  • Antigen symbol:
    CDX2
  • Clonality:
    Monoclonal
  • Clone:
    1C7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    1045
  • Antigen synonyms:
    caudal type homeo box transcription factor 2|homeobox protein CDX-2|CDX3|caudal type homeobox transcription factor 2|CDX-3|caudal-type homeobox protein 2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: CDX2
Clonality: Monoclonal
Clone: 1C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human