Anti-AMPD2 Mouse Monoclonal Antibody [clone: 3C5]
ABNOH00000271-M05
: Abnova
- Antibody type:Primary
- Antigen name:AMP deaminase 2
- Antigen symbol:AMPD2
- Clonality:Monoclonal
- Clone:3C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:271
- Antigen synonyms:PCH9|SPG63
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: AMPD2
Clonality: Monoclonal
Clone: 3C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human