Rat IgG1 Isotype Control [clone: HRPN] (PE (Phycoerythrin)/Cy7®)
Katalog # 60-4301-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:HRPN
- Conjugation:PE (Phycoerythrin)/Cy7®
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.
Type: Control
Antigen: IgG2b (Rat) (rat IgG2b)
Clonality: monoclonal
Clone: HRPN
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: