Mouse IgG2a Isotype Control [clone: C1.18.4] (PE (Phycoerythrin))
50-4724-U100
- Antibody type:Control
- Clonality:Monoclonal
- Clone:C1.18.4
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.
Type: Control
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: monoclonal
Clone: C1.18.4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: