Order Entry
Denmark
ContactUsLinkComponent
Anti-DTX1 Rabbit Polyclonal Antibody
Anti-DTX1 Rabbit Polyclonal Antibody
Catalog # ABGEAP8923C
Supplier:  Abgent
undefined
Anti-DTX1 Rabbit Polyclonal Antibody
Catalog # ABGEAP8923C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Deltex 1
  • Antigen symbol:
    DTX1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Gene ID:
    5150
  • Antigen synonyms:
    hDx-1|Deltex homolog 1
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    RRGSHPYIDFRIFHC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,3 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This PDE7A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 66-94 amino acids from the N-terminal region of human PDE7A.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: DTX1 (deltex 1, E3 ubiquitin ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human