Anti-PP5 (PP5C) Rabbit Polyclonal Antibody
Catalog # ABGEAP8476B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Protein Phosphatase 5, Catalytic Subunit
- Antigen symbol:PP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Gene ID:3792
- Antigen synonyms:FLJ36922|PPT|PP5|PP-T|PPP5C|EC 3.1.3.16|PPP5
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:EFDVPLKQDQEQKIYC
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:This KEL antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 214-243 amino acids from the Central region of human KEL.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PP5 (PP5C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human