Anti-MPST Rabbit Polyclonal Antibody
Catalog # ABGEAP7652B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Mercaptopyruvate sulfurtransferase
- Antigen symbol:MPST
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:TST2|MST|TUM1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MPST (mercaptopyruvate sulfurtransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human