Anti-MOK Rabbit Polyclonal Antibody
Catalog # ABGEAP7543B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:MOK Protein Kinase
- Antigen symbol:MOK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal (350–450 aa)
- Cross adsorption:No
- Antigen synonyms:MAPK/MAK/MRK overlapping kinase|MOK protein kinase|RAGE-1|renal tumor antigen 1|RAGE|renal cell carcinoma antigen|RAGE1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MOK (MOK protein kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human