Order Entry
Denmark
ContactUsLinkComponent
Anti-PRKAG3 Rabbit Polyclonal Antibody
Anti-PRKAG3 Rabbit Polyclonal Antibody
Catalog # ABGEAP7050C
Supplier:  Abgent
undefined
Anti-PRKAG3 Rabbit Polyclonal Antibody
Catalog # ABGEAP7050C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 3
  • Antigen symbol:
    PRKAG3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Center (100–200 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    AMPKG3|AMPK gamma3|gamma-3 subunit|AMPK gamma-3 chain|5'-AMP-activated protein kinase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: PRKAG3 (protein kinase, AMP-activated, gamma 3 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse