Order Entry
Denmark
ContactUsLinkComponent
Anti-KCNJ2 Rabbit Polyclonal Antibody
Anti-KCNJ2 Rabbit Polyclonal Antibody
Catalog # ABGEAP6926C
Supplier:  Abgent
undefined
Anti-KCNJ2 Rabbit Polyclonal Antibody
Catalog # ABGEAP6926C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Potassium Inwardly-rectifying Channel Subfamily J Member 2
  • Antigen symbol:
    KCNJ2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Antigen synonyms:
    ECHD|PBFE|Peroxisomal bifunctional enzyme|3-hydroxyacyl-CoA dehydrogenase|Enoyl-CoA hydratase/3,2-trans-enoyl-CoA isomerase|PBE
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: KCNJ2 (potassium inwardly-rectifying channel, subfamily J, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse