Anti-GALT Rabbit Polyclonal Antibody
Catalog # ABGEAP6880B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Galactose-1-Phosphate Uridylyltransferase
- Antigen symbol:GALT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:UDP glucose hexose 1 phosphate uridylyltransferase|GALT_HUMAN.|Galactose 1 phosphate uridyl transferase|Gal 1 P uridylyltransferase|Galactose 1 phosphate uridylyltransferase
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: GALT (galactose-1-phosphate uridylyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human