Ordreindtastning
Denmark
ContactUsLinkComponent
 
Anti-CLUAP1 Rabbit Polyclonal Antibody
Catalog # ABGEAP6632B
Supplier:  Abgent
undefined
Anti-CLUAP1 Rabbit Polyclonal Antibody
Catalog # ABGEAP6632B
Supplier:  Abgent

Specifications

  • Antistof type:
    Primary
  • Antigen name:
    Seryl-tRNA synthetase 2, mitochondrial
  • Antigen symbol:
    SARS2
  • Clonality:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flowcytometri:
    Yes
  • Vært:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Antigen synonymer:
    EC 6.1.1.11|mitochondrial [Precursor]|Serine--tRNA ligase|Seryl-tRNA synthetase|SerRSmt|SARS2|SARSM|Seryl-tRNA(Ser/Sec) synthetase
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Pakn:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CLUAP1 (clusterin associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse