Anti-CRAT Rabbit Polyclonal Antibody
Catalog # ABGEAP6565A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Carnitine O-acetyltransferase
- Antigen symbol:CRAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:Carnitine O-acetyltransferase|CRAT|CrAT|CAT1|Carnitine acetylase|Carnitine acetyltransferase|CAT
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: CRAT (carnitine O-acetyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human