Anti-BLNK Rabbit Polyclonal Antibody
Katalog # ABGEAP6502C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:B-cell linker
- Antigen symbol:BLNK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:Src homology 2 domain-containing leukocyte protein of 65 kDa|B-cell adapter containing a Src homology 2 domain protein|B cell adaptor containing SH2 domain|B cell linker protein|cytoplasmic adapter protein|SLP-65|B-cell activation|LY57|AGM4|B-cell adapter containing a SH2 domain protein|BLNK-S|bca|B-cell linker protein|SLP65|BASH|Src homology [SH2] domain-containing leukocyte protein of 65 kD
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: BLNK (B-cell linker)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human