Anti-PRDX6 Rabbit Polyclonal Antibody
ABGEAP2927B
: Abgent
- Antibody type:Primary
- Antigen name:Peroxiredoxin 6
- Antigen symbol:PRDX6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:KIAA0106|MGC46173|1-Cys PRX|AOP2|NSGPx|liver 2D page spot 40|peroxiredoxin-6|p29|antioxidant protein 2|1-Cys|PRX|non-selenium glutathione peroxidase|24 kDa protein|red blood cells page spot 12|aiPLA2|1-Cys peroxiredoxin|acidic calcium-independent phospholipase A2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: PRDX6 (peroxiredoxin 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human