Anti-RMND5B Rabbit Polyclonal Antibody
ABGEAP2798B
: Abgent
- Antibody type:Primary
- Antigen name:RMD5 homolog B
- Antigen symbol:RMND5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:RP23-79E13.4|FLJ22318|MGC112682|RMD5B_HUMAN.|Required for meiotic nuclear division 5 homolog B S. cerevisiae |Protein RMD5 homolog B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: RMND5B (required for meiotic nuclear division 5 homolog B (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human