Anti-PPAT Rabbit Polyclonal Antibody
ABGEAP2752C
: Abgent
- Antibody type:Primary
- Antigen name:Phosphoribosyl Pyrophosphate Amidotransferase
- Antigen symbol:PPAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:ATase|EC 2.4.2.14|PPAT|Glutamine phosphoribosylpyrophosphate amidotransferase|GPAT|Amidophosphoribosyltransferase [Precursor]
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: PPAT (phosphoribosyl pyrophosphate amidotransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human