Order Entry
Denmark
ContactUsLinkComponent
Anti-NEGR1 Rabbit Polyclonal Antibody
Anti-NEGR1 Rabbit Polyclonal Antibody
Catalog # ABGEAP16955C
Supplier:  Abgent
undefined
Anti-NEGR1 Rabbit Polyclonal Antibody
Catalog # ABGEAP16955C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neuronal Growth Regulator 1
  • Antigen symbol:
    NEGR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Center
  • Gene ID:
    257194
  • Antigen synonyms:
    DMML2433|IGLON4|Ntra|KILON
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molecular weight:
    38.719 kDa
  • Sequence:
    SAENDVSFPDVRKVKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This NEGR1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 189-218 amino acids from the Central region of human NEGR1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: NEGR1 (neuronal growth regulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human