Anti-CDKL4 Rabbit Polyclonal Antibody
ABGEAP16075B
: Abgent
- Antistof type:Primary
- Antigen name:Cyclin-dependent kinase-like 4
- Antigen symbol:CDKL4
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Epitop:C-Terminal
- Gen ID:381113
- Antigen synonymer:AU067824|Gm942
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:39.446 kDa
- Sekvens:ESFQEDQMKRKARC
- Opbevaringstemperatur:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Koncentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Mouse Cdkl4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 272-299 amino acids from the C-terminal region of mouse Cdkl4.
- Purification:Protein A & Peptide affinity purification
- Pakn:400 µl
Type: Primary
Antigen: CDKL4 (cyclin-dependent kinase-like 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse