Anti-IL33 Rabbit Polyclonal Antibody
ABGEAP14363A
: Abgent
- Antibody type:Primary
- Antigen name:Interleukin 33
- Antigen symbol:IL33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Gene ID:90865
- Antigen synonyms:NFEHEV|C9orf26|IL1F11|NF-HEV|Propeptide|DVS27
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:30.759 kDa
- Sequence:TKRPSLKTGRKHKRHC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This IL33 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 50-79 amino acids from the N-terminal region of human IL33.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: IL33 (interleukin 33)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human