Anti-HOXA6 Rabbit Polyclonal Antibody
ABGEAP12025C
: Abgent
- Antibody type:Primary
- Antigen name:Homeobox A6
- Antigen symbol:HOXA6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:homeo box 1B|homeobox protein Hox-A6|homeobox protein Hox-1B|HOX1B|HOX1.2|HOX1|homeobox protein HOXA6|homeo box A6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: HOXA6 (homeobox A6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human