Ordreindtastning
Denmark
ContactUsLinkComponent
Anti-CASKIN2 Rabbit Polyclonal Antibody
Anti-CASKIN2 Rabbit Polyclonal Antibody
Catalog # ABGEAP10948C
Leverandør:  Abgent
Anti-CASKIN2 Rabbit Polyclonal Antibody
Catalog # ABGEAP10948C
Leverandør:  Abgent

Specifikationer

  • Antistof type:
    Primary
  • Antigen name:
    CASK Interacting Protein 2
  • Antigen symbol:
    CASKIN2
  • Clonality:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Vært:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-Terminal
  • Krydsadsorption:
    No
  • Antigen synonymer:
    KIAA1139|ANKS5B|Caskin-2|CASK-interacting protein 2|CASKIN2
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Pakn:
    400 µl

Specifikationer

Om denne vare

Type: Primary
Antigen: CASKIN2 (CASK interacting protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human