Anti-PRKAA2 Rabbit Polyclonal Antibody
Catalog # ABGEAP10846A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase AMP-activated Catalytic Subunit Alpha 2
- Antigen symbol:PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (1271–1288 aa)
- Cross adsorption:No
- Antigen synonyms:AMPKa2|AMPK|AMPK2|PRKAA
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PRKAA2 (protein kinase, AMP-activated, alpha 2 catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human