Anti-CES1P1 Rabbit Polyclonal Antibody
Catalog # ABGEAP10710A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase AMP-activated Catalytic Subunit Alpha 2
- Antigen symbol:PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (450–550 aa)
- Cross adsorption:No
- Antigen synonyms:AMPKa2|AMPK|AMPK2|PRKAA
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: CES1P1 (carboxylesterase 1 pseudogene 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human