Anti-PURG Rabbit Polyclonal Antibody
ABGEAP10617B
: Abgent
- Antibody type:Primary
- Antigen name:Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 1
- Antigen symbol:PRKAG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (200–300 aa)
- Cross adsorption:No
- Antigen synonyms:5'-AMP-activated protein kinase subunit gamma-1|AMPKG3|AMPK gamma3|PRKAG3|AMPK- gamma3|5'-AMP-activated protein kinase subunit gamma-2|AMPK- gamma1|AMPK subunit gamma-1|AMPKg|AMPK subunit gamma-3|H91620p|PRKAG2|AMPK- gamma2|AMPK subunit gamma-2|PRKAG1|AMPK gamma2|AMPK gamma1|5'-AMP-activated protein kinase subunit gamma-3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: PURG (purine-rich element binding protein G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human