Order Entry
Denmark
ContactUsLinkComponent
 
Anti-HFM1 Rabbit Polyclonal Antibody
Catalog # ABGEAP10614B
Supplier:  Abgent
Anti-HFM1 Rabbit Polyclonal Antibody
Catalog # ABGEAP10614B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HFM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal (1–100 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    kinase AMPK-beta1|AMPK-beta1|AMPK beta-1 chain|5'-AMP-activated protein kinase beta-1 subunit|PRKAB1|AAKB1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: HFM1 (HFM1, ATP-dependent DNA helicase homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human