Anti-CCNYL2 Rabbit Polyclonal Antibody
Catalog # AVIVARP69195_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:cyclin Y-like 2
- Antigen symbol:CCNYL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CCNYL2 (cyclin Y-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human