Anti-LAMC3 Rabbit Polyclonal Antibody
AVIVARP65777_P050
- Antibody type:Primary
- Antigen name:laminin, gamma 3
- Antigen symbol:LAMC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10319
- Antigen synonyms:OCCM
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: LAMC3 (laminin, gamma 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human