Anti-NOS2 Rabbit Polyclonal Antibody
Katalog # AVIVARP63000_P050
Leverandør: Aviva Systems Biology
Specifikationer
- Antistof type:Primary
- Antigen name:Nitric Oxide Synthase 2
- Antigen symbol:NOS2
- Clonality:Polyclonal
- Konjugation:Unconjugated
- Vært:Rabbit
- Isotype:
- Reactivity:Pig,Human
- Western blot:Yes
- Environmentally Preferable:
- Epitop:N-terminal
- Krydsadsorption:none
- Gen ID:4843
- Antigen synonymer:INOS|HEP-NOS|NOS2A|NOS
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Pakn:50 µG
Specifikationer
Om denne vare
Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Pig