Anti-KIAA0226 Rabbit Polyclonal Antibody
Catalog # AVIVARP58950_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen symbol:KIAA0226
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:9711
- Antigen synonyms:SCAR15|RUBICON
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: KIAA0226 (KIAA0226)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: