Anti-LGALS4 Rabbit Polyclonal Antibody
Catalog # AVIVARP42280_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:lectin, galactose binding, soluble 4
- Antigen symbol:LGALS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:16855
- Antigen synonyms:gal-4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: LGALS4 (lectin, galactoside-binding, soluble, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: