Anti-CSF2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3790R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Colony Stimulating Factor 2
- Antigen symbol:CSF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:GMCSF|TUBB2|Csfgm|Gm-CSf|TUBB|MGI-IGM|CDCBM5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: