Anti-CYP2D6 Rabbit Polyclonal Antibody
Katalog # BOSSBS-2991R
Leverandør: Bioss
Specifikationer
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 2, Subfamily D, Polypeptide 6
- Antigen symbol:CYP2D6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:P450C2D|CYP2D7BP|P450-DB1|CYP2DL1|P450DB1|CYP2D7AP|Cytochrome P450 2D6|CYP2D8P2|CYP2D7P2|CYPIID6|CYP2D|CPD6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifikationer
Om denne vare
Type: Primary
Antigen: CYP2D6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: