Anti-CACNA1G Rabbit Polyclonal Antibody
Catalog # BOSSBS-2781R
Supplier: Bioss
Specifications
- Antistof type:Primary
- Antigen name:Calcium Channel, Voltage-dependent, T type, alpha 1G subunit
- Antigen symbol:CACNA1G
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Rabbit
- ImmunoChemistry:Yes
- ImmunFluorescens:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Krydsadsorption:none
- Antigen synonymer:Cav3.1d|[a]1G|mKIAA1123|a1G|alpha-1G
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Pakn:100 µl
Specifications
About this item
Type: Primary
Antigen: CACNA1G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat