Anti-AR Rabbit Polyclonal Antibody
BOSSBS-12471R
: Bioss
- Antistof type:Primary
- Antigen name:Androgen Receptor
- Antigen symbol:AR
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Rabbit
- ImmunoChemistry:Yes
- ImmunFluorescens:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Krydsadsorption:none
- Antigen synonymer:AR|Nuclear receptor subfamily 3 group C member 4|NR3C4|Dihydrotestosterone receptor|DHTR
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Pakn:100 µl
Type: Primary
Antigen: AR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human