Anti-FGA Rabbit Polyclonal Antibody
Catalog # BOSSBS-1240R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Fibrinogen
- Antigen symbol:FGA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:FGA|Leucine-rich repeats and calponin homology (CH) Domain containing 4|Fibrinogen alpha chain|Fibrinogen A alpha polypeptide chain|Fib2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: human Fibrinogen
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human