Anti-CADM1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1147R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cell adhesion molecule 1
- Antigen symbol:CADM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:synCAM1|IGSF4|sgIGSF|SYNCAM|BL2|IGSF4A|TSLC1|NECL2|ST17|Necl-2|sTSLC-1|RA175
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CADM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat