Anti-PDGFA Rabbit Polyclonal Antibody
BOSSBS-0196R
: Bioss
- Antibody type:Primary
- Antigen name:Platelet-derived growth factor alpha
- Antigen symbol:PDGFA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:Platelet-derived growth factor alpha polypeptide|PDGFA|Platelet derived growth factor A chain precursor|PDGF1|Platelet-derived growth factor A chain|PlateletΓÇôdirived growth factorΓÇô1|Platelet derived growth factor alpha polypeptide|PDGF-1|Platelet-derived growth factor subunit A.|PDGFA_HUMAN|PDGF AA|PDGF A chain|PDGF subunit A|PDGF-AA
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: PDGFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat