Order Entry
Denmark
ContactUsLinkComponent
Trays for Neptune® pipette tips
Trays for Neptune® pipette tips
Catalog # BITX2162.T
Supplier:  BIOTIX
Trays for Neptune® pipette tips
Catalog # BITX2162.T
Supplier:  BIOTIX
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Tray with insert card, for bulk tips and ESP reload
  • For use with:
    1000 μl tips
  • Pk:
    1
Specifications
About this item
Manufactured from recycled polypropylene and developed to use the least amount of plastic.
  • Recyclable tray
  • Less waste
  • Maximum space saving design
These trays reduce plastic waste by up to 90% over conventional trays in combination with the ESP reload system.