- Description:Biosafety cabinets, Purifier® Logic®, class II, type B2, nominal width 6 feet
- Environmentally Preferable:Energy Efficient,Sustainable Materials
- Model:Purifier® Logic® - 6 feet
- Electrical connection:230 V, 50/60 Hz, 8 A, UK plug
- Exhaust air volume:1697 m³/h
- Ext. W×D×H:1989×792×2543 mm
- Weight:426,4 kg
- Pk:1
Purifier Logic+ Class II, Type B2 biosafety cabinets provides personnel, product and environmental protection from hazardous particulates such as agents that require Biosafety Level 1, 2 or 3 containment. Other appropriate applications include work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates.
- MyLogic provides intuitive programming colour coded menus, status messages and alarms with diagnostic troubleshooting-all using full sentences
- 10° angled sash
- Patented curved air grille with built in armrest
- Fully accessible controls
- Eye-level mounted colour display
Using the same ECM blower technology as the Purifier Logic®, the Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
These biosafety cabinets are designed to be hard ducted to the outside. The cabinet provides personnel, product and environmental protection and is suitable for work in biosafety levels 1, 2 and 3. In addition, some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included.