Order Entry
Denmark
ContactUsLinkComponent
Bulb pipettes, one mark, Class A
Bulb pipettes, one mark, Class A
Catalog # HIRS134G15
Supplier:  HIRSCHMANN
Bulb pipettes, one mark, Class A
Catalog # HIRS134G15
Supplier:  HIRSCHMANN
Specifications
  • Capacity:
    15 ml
  • Description:
    Unserialised pipettes
  • Seq:
    03
  • Pk:
    6
  • Tolerance:
    0,03 ± ml
Specifications
About this item
Borosilicate glass.
  • Designed to deliver according to ASTM Specification E969, Class A requirements
  • Colour-coded for ease in sorting and selection
  • Fine black enamel capacity line and markings
  • Serialised pipettes include a Certificate of Identification and Accuracy
Certifications: ASTM E969