VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Certificates
About this item
Cardboard, coated.
- Waterproof coating
- Various combinations possible with different dividers (to be ordered separately)
- Available in different heights and colours
For storage of cryovials at low temperatures.
Freezable to –140 °C.
Frequently Bought Together