Order Entry
Denmark
ContactUsLinkComponent
SP Bel-Art® liquid sampler
SP Bel-Art® liquid sampler
Catalog # BELAH378270000
undefined
SP Bel-Art® liquid sampler
Catalog # BELAH378270000
Specifications
  • Capacity:
    50 ml
  • Description:
    Liquid sampler
  • Length:
    500 mm
  • Material:
    Nickel plated brass
  • Pk:
    1
Specifications
About this item
Liquid sampler made of nickel-plated brass cup and tubular handle with finger grip. Cup is not replaceable and can not detach.
Suitable for immersion into a can, bottle, tank, vat ...