Order Entry
Denmark
ContactUsLinkComponent
Bovine deoxyribonuclease I (from Pancreas)
Bovine deoxyribonuclease I (from Pancreas)
Catalog # ROCKMB-101-0100
CAS Number:  
Bovine deoxyribonuclease I (from Pancreas)
Catalog # ROCKMB-101-0100
CAS Number:  

Specifications

  • Pk:
    0,1 KU
  • Source:
    Pancreas
  • Species:
    Bovine
  • Enzyme name:
    Deoxyribonuclease I

Specifications

About this item

Deoxyribonuclease I is an endonuclease which splits phosphodiester linkages yielding polynucleotides with a free 3'-OH and 5'-phosphate. Activated by bivalent metal ions and inhibited by chelators such as EDTA. Chromatographically purified to be free of ribonuclease and protease activity.