Order Entry
Denmark
Orders LinkContactUsLinkComponent
ECR masking reagent solution
ECR masking reagent solution
ECR masking reagent solutionfor aluminium determination, 0,006 - 0,25 mg/l, Eriochrome cyanine R, 25 mL
Catalog # HACH2380123
Supplier:   Hach
ECR masking reagent solution
Catalog # HACH2380123
Supplier:  Hach
CAS Number:  
ECR masking reagent solution
ECR masking reagent solution
ECR masking reagent solutionfor aluminium determination, 0,006 - 0,25 mg/l, Eriochrome cyanine R, 25 mL
CAS Number:  
Price
248,00 kr
Log in for availability
1 * 25 mL
Product Details & Documents
This solution is used for Aluminum determination by the Eriochrome Cyanine R Method.
Documents
Specifications
Specifications
  • Chemical method:
    Eriochrome cyanine R
  • Pk:
    25 mL
  • Description:
    ECR masking reagent solutionfor aluminium determination
  • Range:
    0,006 - 0,25 mg/l