Order Entry
Denmark
Orders LinkContactUsLinkComponent
Visual colour kits, glutaraldehyde
Visual colour kits, glutaraldehyde
Glutaraldehyde test kit, model GT-1, colour disc, 0,5 - 2800 mg/l, 1 KIT
Catalog # HACH2587200
Supplier:   Hach
Visual colour kits, glutaraldehyde
Catalog # HACH2587200
Supplier:  Hach
CAS Number:  
Visual colour kits, glutaraldehyde
Visual colour kits, glutaraldehyde
Glutaraldehyde test kit, model GT-1, colour disc, 0,5 - 2800 mg/l, 1 KIT
CAS Number:  
Price
6.780,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
Glutaraldehyde test kits are designed to monitor disinfection processes.
  • Simple, economical colour disc method
  • Accurate and quick results
  • Easy-to-carry test kit
Delivery information: Kits includes three reagents, colour discs, colour comparator box, bottles, sprinkler caps, syringes, cylinders, marker, ampoule breaker, ultra pure water, viewing tubes, instruction sheet, and carrying case.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Glutaraldehyde test kit, model GT-1, colour disc
  • Range:
    0,5 - 2800 mg/l