Sodium chloride 26% in aqueous solution, VOLUSOL®
6241.9011
:
- Pack type:Bag-in-box
- Pk:10 L
- Seq:5214
- Environmentally Preferable:
: Saline, NaCl, Sea salt, Halite
:
:58.44 g/mol
:1.195 g/cm³ (20 °C)
:Ambient
:231-598-3
:7647-14-5
:NaCl
:MFCD00003477