Multiparameter test kits, model HA-62
Supplier: Hach
Jump to:
About this item
A multi-parameter test kit for hardness, iron and pH, which provides an easy-to-use, cost effective way to determine the quality of water.
- Popular tests in one rugged kit
- Uses color disc and drop count titration methods
2 Options Available Below
Product Details & Documents
A multi-parameter test kit for hardness, iron and pH, which provides an easy-to-use, cost effective way to determine the quality of water.
- Popular tests in one rugged kit
- Uses color disc and drop count titration methods
Product Family Options
Product Information
- RangePkAvailabilityPrice
- HACH183700For hardness (as CaCO₃), iron and pH, HA-62For hardness (as CaCO₃), iron and pH, HA-62Hardness: 17,19 - 342,36 mg/l, Iron: 0,1 - 5 mg/l, pH: 6,5 - 8,51 KITLog in for availability1 * 1 Each3.470,00 kr
- HACH183702For hardness (as CaCO₃), iron and pH, HA-62BFor hardness (as CaCO₃), iron and pH, HA-62BHardness: 17,19 - 342,36 mg/l, Iron: 0,2 - 10 mg/l, pH: 4 - 101 KITLog in for availability1 * 1 Each3.880,00 kr