Order Entry
Denmark
ContactUsLinkComponent
Cleanroom wipes, pre-saturated, sterile AlphaSat® 10
Cleanroom wipes, pre-saturated, sterile AlphaSat® 10
Catalog # 115-1891
Supplier:  Texwipe
undefined
Cleanroom wipes, pre-saturated, sterile AlphaSat® 10
Catalog # 115-1891
Supplier:  Texwipe
Specifications
  • Packed:
    20 wipes/flexpack; 20 packs/case
  • Description:
    TX3285
  • L×W:
    230×230 mm
  • Pk:
    400
Specifications
About this item
This 100% continuous filament polyester wipe with thermally sealed borders is engineered to have the lowest level of releasable particles and fibres for critical and sterile environments. The wipe is pre-wetted with 70% USP IPA.
  • Optimises IPA use for effective particulate removal and eliminates overspray of alcohol
  • Superior strength for rigorous wiping without snagging on equipment
  • Wipe glides smoothly over surfaces with low abrasion
  • Sized appropriately for limited isolator space
Ideal for cleaning cleanroom environments including the interiors of Compounding Aseptic Isolators (CAIs), aseptic isolators in pharmaceutical manufacturing, and Biological Safety Cabinets (BSCs).
Suitable for use in Grade A, ISO Class 3 - 5 cleanroom environments.
Packaging: Double bagged in a small resealable flexpack package for easy entry into aseptic and sterile isolators.